3-Bromo-4-hydroxy-6-trifluoromethylquinoline

Lieferant: Apollo Scientific

PC404592-1G
APOSPC404592-1GEA 556 EUR
APOSPC404592-1G
3-Bromo-4-hydroxy-6-trifluoromethylquinoline
3-Bromo-4-hydroxy-6-trifluoromethylquinoline
Formel: C₁₀H₅BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD13192876
CAS-Nummer: 1065087-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR