4-Hydroxy-7-trifluoromethoxyquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

PC404593-1G
APOSPC404593-1GEA 525 EUR
APOSPC404593-1G
4-Hydroxy-7-trifluoromethoxyquinoline-3-carboxylic acid
4-Hydroxy-7-trifluoromethoxyquinoline-3-carboxylic acid
Formel: C₁₁H₆F₃NO₄
Molecular Weight: 273,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192937
CAS-Nummer: 53985-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR