3-Brom-4-chlor-6-fluorchinolin

Lieferant: Apollo Scientific

PC404594-1G
APOSPC404594-1GEA 482 EUR
APOSPC404594-1G
3-Brom-4-chlor-6-fluorchinolin
3-Bromo-4-chloro-6-fluoroquinoline
Formel: C₉H₄BrClFN
Molecular Weight: 260,4900 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192958
CAS-Nummer: 1204810-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR