3-Bromo-4-chloro-7-fluoroquinoline
Lieferant: Apollo Scientific
PC404596-1G
APOSPC404596-1GEA
475
EUR
APOSPC404596-1G
3-Bromo-4-chloro-7-fluoroquinoline
3-Bromo-4-chloro-7-fluoroquinoline
Formel:
C₉H₄BrClFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD13192960 CAS-Nummer: 1204810-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...