3,4-Dichloro-7-fluoroquinoline

Lieferant: Apollo Scientific

PC404597-1G
APOSPC404597-1GEA 491 EUR
APOSPC404597-1G
3,4-Dichloro-7-fluoroquinoline
3,4-Dichloro-7-fluoroquinoline
Formel: C₉H₄Cl₂FN
Molecular Weight: 216,039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192961
CAS-Nummer: 1204810-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR