3,4-Dichloro-8-fluoroquinoline

Lieferant: Apollo Scientific
Achtung

PC404598-1G
APOSPC404598-1GEA 393 EUR
APOSPC404598-1G
3,4-Dichloro-8-fluoroquinoline
3,4-Dichloro-8-fluoroquinoline
Formel: C₉H₄Cl₂FN
Molecular Weight: 216,039 g/mol
Schmelzpunkt: 107…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13192963
CAS-Nummer: 1204811-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR