3-Bromo-4-chloro-6-trifluoromethylquinoline

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-4-chloro-6-(trifluoromethyl)quinoline

PC404599-1G
APOSPC404599-1GEA 74.1 EUR
APOSPC404599-1G
3-Bromo-4-chloro-6-trifluoromethylquinoline
3-Bromo-4-chloro-6-trifluoromethylquinoline
Formel: C₁₀H₄BrClF₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD13192972
CAS-Nummer: 1204810-99-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR