3,4-Dichloro-5,7-difluoroquinoline

Lieferant: Apollo Scientific

PC404600-1G
APOSPC404600-1GEA 475 EUR
APOSPC404600-1G
3,4-Dichloro-5,7-difluoroquinoline
3,4-Dichloro-5,7-difluoroquinoline
Formel: C₉H₃Cl₂F₂N
Molecular Weight: 234.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192995
CAS-Nummer: 1204810-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR