3,4-Dichloro-5,8-difluoroquinoline

Lieferant: Apollo Scientific

PC404601-1G
APOSPC404601-1GEA 475 EUR
APOSPC404601-1G
3,4-Dichloro-5,8-difluoroquinoline
3,4-Dichloro-5,8-difluoroquinoline
Formel: C₉H₃Cl₂F₂N
Molecular Weight: 234,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192997
CAS-Nummer: 1204811-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR