4-Bromo-3-chloro-6-fluoroquinoline

Lieferant: Apollo Scientific

PC404602-1G
APOSPC404602-1GEA 542 EUR
APOSPC404602-1G
4-Bromo-3-chloro-6-fluoroquinoline
4-Bromo-3-chloro-6-fluoroquinoline
Formel: C₉H₄BrClFN
Molecular Weight: 260,49 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619698
CAS-Nummer: 724787-81-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR