4-Bromo-3-chloro-8-fluoroquinoline

Lieferant: Apollo Scientific

PC404604-1G
APOSPC404604-1GEA 542 EUR
APOSPC404604-1G
4-Bromo-3-chloro-8-fluoroquinoline
4-Bromo-3-chloro-8-fluoroquinoline
Formel: C₉H₄BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD13619702
CAS-Nummer: 1211659-63-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR