4-Bromo-3-chloro-5,7-difluoroquinoline

Lieferant: Apollo Scientific

PC404605-1G
APOSPC404605-1GEA 614 EUR
APOSPC404605-1G
4-Bromo-3-chloro-5,7-difluoroquinoline
4-Bromo-3-chloro-5,7-difluoroquinoline
Formel: C₉H₃BrClF₂N
Molecular Weight: 278,481 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619734
CAS-Nummer: 1209122-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR