3,4-Dibromo-6,8-difluoroquinoline
Lieferant: Apollo Scientific
PC404607-1G
APOSPC404607-1GEA
556
EUR
APOSPC404607-1G
3,4-Dibromo-6,8-difluoroquinoline
3,4-Dibromo-6,8-difluoroquinoline
Formel:
C₉H₃Br₂F₂N Molecular Weight: 322,932 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13619737 CAS-Nummer: 1210168-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...