4-Chloro-7-(trifluoromethoxy)quinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC404610-1G
APOSPC404610-1GEA 672 EUR
APOSPC404610-1G
4-Chloro-7-(trifluoromethoxy)quinoline-3-carboxylic acid ethyl ester
4-Chloro-7-(trifluoromethoxy)quinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₉ClF₃NO₃
Molecular Weight: 319.664 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987428
CAS-Nummer: 890841-23-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR