4-Chloro-8-(trifluoromethoxy)quinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC404611-1G
APOSPC404611-1GEA 509 EUR
APOSPC404611-1G
4-Chloro-8-(trifluoromethoxy)quinoline-3-carboxylic acid ethyl ester
4-Chloro-8-(trifluoromethoxy)quinoline-3-carboxylic acid ethyl ester
Formel: C₁₃H₉ClF₃NO₃
Molecular Weight: 319,664 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13366286
CAS-Nummer: 1040013-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR