4-Bromo-6-fluoroquinoline-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC404612-1G
APOSPC404612-1GEA 509 EUR
APOSPC404612-1G
4-Bromo-6-fluoroquinoline-3-carboxylic acid ethyl ester
4-Bromo-6-fluoroquinoline-3-carboxylic acid ethyl ester
Formel: C₁₂H₉BrFNO₂
Molecular Weight: 298.108 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987487
CAS-Nummer: 1242260-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR