8-Fluoro-2-methylquinoline-3-carboxylic acid ethyl ester
Lieferant: Apollo Scientific
PC404615-1G
APOSPC404615-1GEA
0
EUR
APOSPC404615-1G
8-Fluoro-2-methylquinoline-3-carboxylic acid ethyl ester
8-Fluoro-2-methylquinoline-3-carboxylic acid ethyl ester
Formel:
C₁₃H₁₂FNO₂ Molecular Weight: 233,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20036311 CAS-Nummer: 1333253-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...