4-Bromo-3-chloro-5,8-difluoro-2-methylquinoline
Lieferant: Apollo Scientific
PC404616-1G
APOSPC404616-1GEA
525
EUR
APOSPC404616-1G
4-Bromo-3-chloro-5,8-difluoro-2-methylquinoline
4-Bromo-3-chloro-5,8-difluoro-2-methylquinoline
Formel:
C₁₀H₅BrClF₂N Molecular Weight: 292,507 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20036486 CAS-Nummer: 1333252-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...