6-Fluoro-4-(piperazin-1-yl)quinoline Hydrochloride

Lieferant: Apollo Scientific

PC404617-1G
APOSPC404617-1GEA 383 EUR
APOSPC404617-1G
6-Fluoro-4-(piperazin-1-yl)quinoline Hydrochloride
6-Fluoro-4-(piperazin-1-yl)quinoline Hydrochloride
Formel: C₁₃H₁₄FN₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20036730
CAS-Nummer: 1333249-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR