7-Fluoro-4-(piperazin-1-yl)quinoline Hydrochloride

Lieferant: Apollo Scientific

PC404618-1G
APOSPC404618-1GEA 0 EUR
APOSPC404618-1G
7-Fluoro-4-(piperazin-1-yl)quinoline Hydrochloride
7-Fluoro-4-(piperazin-1-yl)quinoline Hydrochloride
Formel: C₁₃H₁₄FN₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20036731
CAS-Nummer: 1333252-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR