2-Chlor-6-fluor-4-methylchinolin
Lieferant: Apollo Scientific
PC404620-1G
APOSPC404620-1GEA
201
EUR
APOSPC404620-1G
2-Chlor-6-fluor-4-methylchinolin
2-Chloro-6-fluoro-4-methylquinoline
Formel:
C₁₀H₇ClFN Lagertemperatur: Raumtemperatur |
CAS-Nummer:
18529-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...