2-Chloro-7-fluoroquinoline

Lieferant: Apollo Scientific

PC404622-1G
APOSPC404622-1GEA 0 EUR
APOSPC404622-1G
2-Chloro-7-fluoroquinoline
2-Chloro-7-fluoroquinoline
Formel: C₉H₅ClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD09261127
CAS-Nummer: 445041-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR