Ethyl 4-chloro-7-fluoroquinazoline-2-carboxylate
Lieferant: Apollo Scientific
PC404623-1G
APOSPC404623-1GEA
317
EUR
APOSPC404623-1G
Ethyl 4-chloro-7-fluoroquinazoline-2-carboxylate
Ethyl 4-chloro-7-fluoroquinazoline-2-carboxylate
Formel:
C₁₁H₈ClFN₂O₂ Molecular Weight: 254,645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12675148 CAS-Nummer: 1189106-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...