5-Chloro-3-{[2-(trifluoromethyl)phenyl]methyl}-1,2,4-thiadiazole

Lieferant: Apollo Scientific

PC404624-1G
APOSPC404624-1GEA 517 EUR
APOSPC404624-1G
5-Chloro-3-{[2-(trifluoromethyl)phenyl]methyl}-1,2,4-thiadiazole
5-Chloro-3-{[2-(trifluoromethyl)phenyl]methyl}-1,2,4-thiadiazole
Formel: C₁₀H₆ClF₃N₂S
Molecular Weight: 278,681 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20257106
CAS-Nummer: 1338218-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR