(S)-4-(oxiran-2-yl)-2,8-bis(trifluoromethyl)quinoline
Lieferant: Apollo Scientific
PC404626-1G
APOSPC404626-1GEA
0
EUR
APOSPC404626-1G
(S)-4-(oxiran-2-yl)-2,8-bis(trifluoromethyl)quinoline
(S)-4-(oxiran-2-yl)-2,8-bis(trifluoromethyl)quinoline
Formel:
C₁₃H₇F₆NO Molecular Weight: 307,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28580130 CAS-Nummer: 1292282-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...