3-Fluorocyclobutanecarboxylic acid

Lieferant: Apollo Scientific
Achtung

PC405004-250MG
APOSPC405004-250MGEA 423 EUR
APOSPC405004-250MG
3-Fluorocyclobutanecarboxylic acid
3-Fluorocyclobutanecarboxylic acid
Formel: C₅H₇FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09834117
CAS-Nummer: 122665-96-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR