6,6-Difluoro-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 6,6-difluoro-2-azaspiro[3.3]heptane-2-carboxylate

PC405005-250MG
APOSPC405005-250MGEA 633 EUR
APOSPC405005-250MG
6,6-Difluoro-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester
6,6-Difluoro-2-azaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester
Formel: C₁₁H₁₇F₂NO₂
Molecular Weight: 233,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432776
CAS-Nummer: 1264635-66-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR