2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate
Lieferant: Apollo Scientific
|
Achtung
|
PC405006-250MG
APOSPC405006-250MGEA
546
EUR
APOSPC405006-250MG
2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate
2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate
Formel:
C₇H₁₁NO₂·CF₃CO₂H Molecular Weight: 255,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19982773 CAS-Nummer: 1523572-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...