3-(4-Fluorophenyl)oxetan-3-ol
Lieferant: Apollo Scientific
PC405501-250MG
APOSPC405501-250MGEA
632
EUR
APOSPC405501-250MG
3-(4-Fluorophenyl)oxetan-3-ol
3-(4-Fluorophenyl)oxetan-3-ol
Formel:
C₉H₉FO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22370092 CAS-Nummer: 1379812-16-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...