3-[4-(Trifluoromethyl)phenyl]oxetan-3-ol

Lieferant: Apollo Scientific
Achtung

PC405502-250MG
APOSPC405502-250MGEA 655 EUR
APOSPC405502-250MG
3-[4-(Trifluoromethyl)phenyl]oxetan-3-ol
3-[4-(Trifluoromethyl)phenyl]oxetan-3-ol
Formel: C₁₀H₉F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27664912
CAS-Nummer: 26755-29-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR