3-Fluorocyclobutanamine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
PC405503-250MG
APOSPC405503-250MGEA
0
EUR
APOSPC405503-250MG
3-Fluorocyclobutanamine hydrochloride
3-Fluorocyclobutanamine hydrochloride
Formel:
C₄H₈FN·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD21642364 CAS-Nummer: 1284245-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...