1-(Azetidin-3-yl)-3-fluoroazetidine

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-1,3'-biazetidine

PC405510-250MG
APOSPC405510-250MGEA 748 EUR
APOSPC405510-250MG
1-(Azetidin-3-yl)-3-fluoroazetidine
1-(Azetidin-3-yl)-3-fluoroazetidine
Formel: C₆H₁₁FN₂
Molecular Weight: 130,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123282
CAS-Nummer: 1257293-80-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR