1-(Trifluoromethyl)-2-oxa-6-azaspiro[3.3]heptane hemioxalate

Lieferant: Apollo Scientific
Achtung

PC405520-250MG
APOSPC405520-250MGEA 1300 EUR
APOSPC405520-250MG
1-(Trifluoromethyl)-2-oxa-6-azaspiro[3.3]heptane hemioxalate
1-(Trifluoromethyl)-2-oxa-6-azaspiro[3.3]heptane hemioxalate
Lagertemperatur: Gefrierschrank
MDL: MFCD28009690
CAS-Nummer: 1454909-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR