1-(Trifluoromethyl)-2-thia-6-azaspiro[3.3]heptane hemioxalate

Lieferant: Apollo Scientific

PC405521-250MG
APOSPC405521-250MGEA 1300 EUR
APOSPC405521-250MG
1-(Trifluoromethyl)-2-thia-6-azaspiro[3.3]heptane hemioxalate
1-(Trifluoromethyl)-2-thia-6-azaspiro[3.3]heptane hemioxalate
Lagertemperatur: Raumtemperatur
MDL: MFCD28009691
CAS-Nummer: 1980054-87-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR