1-Methoxy-4-(pentafluorosulphanyl)benzene
Lieferant: Apollo Scientific
PC405523-1G
APOSPC405523-1GEA
669
EUR
APOSPC405523-1G
1-Methoxy-4-(pentafluorosulphanyl)benzene
1-Methoxy-4-(pentafluorosulphanyl)benzene
Formel:
C₇H₇F₅OS Molecular Weight: 234,187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16660964 CAS-Nummer: 852469-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...