1-Methoxy-3-(pentafluorosulphanyl)benzene
Lieferant: Apollo Scientific
PC405524-1G
APOSPC405524-1GEA
748
EUR
APOSPC405524-1G
1-Methoxy-3-(pentafluorosulphanyl)benzene
1-Methoxy-3-(pentafluorosulphanyl)benzene
Formel:
C₇H₇F₅OS Lagertemperatur: Raumtemperatur |
MDL:
MFCD22370099 CAS-Nummer: 1272542-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...