(2-Nitro-5-(pentafluorosulphanyl)phenyl)acetonitrile

Lieferant: Apollo Scientific

PC405562-1G
APOSPC405562-1GEA 715 EUR
APOSPC405562-1G
(2-Nitro-5-(pentafluorosulphanyl)phenyl)acetonitrile
(2-Nitro-5-(pentafluorosulphanyl)phenyl)acetonitrile
Formel: C₈H₅F₅N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD22370114
CAS-Nummer: 1309569-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR