(2-Nitro-4-(pentafluorosulphanyl)phenyl)acetonitrile

Lieferant: Apollo Scientific
Achtung

PC405563-1G
APOSPC405563-1GEA 715 EUR
APOSPC405563-1G
(2-Nitro-4-(pentafluorosulphanyl)phenyl)acetonitrile
(2-Nitro-4-(pentafluorosulphanyl)phenyl)acetonitrile
Formel: C₈H₅F₅N₂O₂S
Molecular Weight: 288,195 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22370113
CAS-Nummer: 1309569-40-1
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR