Ethyl (2-nitro-5-(pentafluorosulphanyl)phenyl) acetate

Lieferant: Apollo Scientific

PC405564-1G
APOSPC405564-1GEA 715 EUR
APOSPC405564-1G
Ethyl (2-nitro-5-(pentafluorosulphanyl)phenyl) acetate
Ethyl (2-nitro-5-(pentafluorosulphanyl)phenyl) acetate
Formel: C₁₀H₁₀F₅NO₄S
Molecular Weight: 335,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22370090
CAS-Nummer: 1309569-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR