Ethyl (2-nitro-4-(pentafluorosulphanyl)phenyl) acetate

Lieferant: Apollo Scientific

PC405565-1G
APOSPC405565-1GEA 715 EUR
APOSPC405565-1G
Ethyl (2-nitro-4-(pentafluorosulphanyl)phenyl) acetate
Ethyl (2-nitro-4-(pentafluorosulphanyl)phenyl) acetate
Formel: C₁₀H₁₀F₅NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD22370089
CAS-Nummer: 1394319-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR