2-Brom-4-(pentafluorthio)anilin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-3-bromophenylsulphur pentafluoride, (4-Amino-3-bromophenyl)pentafluorosulphur

PC405596-1G PC405596-250MG PC405596-5G
APOSPC405596-1GEA 345 EUR
APOSPC405596-1G APOSPC405596-250MG APOSPC405596-5G
2-Brom-4-(pentafluorthio)anilin 95%
2-Brom-4-(pentafluorthio)anilin
Formel: C₆H₅BrF₅NS
Molecular Weight: 298.716 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20731078
CAS-Nummer: 159727-25-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR