cis-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid 93%

Lieferant: Apollo Scientific
Gefahr

PC405624-100MG
APOSPC405624-100MGEA 1080 EUR
APOSPC405624-100MG
cis-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid 93%
cis-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid
Formel: C₅H₅F₃O₂
Molecular Weight: 154,087 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17977173
CAS-Nummer: 1258652-18-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR