trans-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

PC405626-100MG
APOSPC405626-100MGEA 715 EUR
APOSPC405626-100MG
trans-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid
trans-2-(Trifluoromethyl)cyclopropane-1-carboxylic acid
Formel: C₅H₅F₃O₂
Molecular Weight: 154,087 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18027181
CAS-Nummer: 78376-99-5
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR