4-(Pentafluorothio)benzylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Aminomethyl)phenylsulphur pentafluoride

PC405655-1G PC405655-5G
APOSPC405655-1GEA 116 EUR
APOSPC405655-1G APOSPC405655-5G
4-(Pentafluorothio)benzylamine
4-(Pentafluorothio)benzylamine
Formel: C₇H₈F₅NS
Molecular Weight: 233.20 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06213675
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR