4-(Pentafluorothio)benzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
PC405656-5G
PC405656-1G
PC405656-500MG
APOSPC405656-5GEA
0
EUR
APOSPC405656-5G
APOSPC405656-1G
APOSPC405656-500MG
4-(Pentafluorothio)benzyl bromide
4-(Pentafluorothio)benzyl bromide
Formel:
C₇H₆BrF₅S Schmelzpunkt: 42…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652420 CAS-Nummer: 1126969-29-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...