3-Bromo-5-(pentafluorothio)benzyl bromide 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Bromo-5-(pentafluorosulphur)benzyl bromide

PC405658-1G
APOSPC405658-1GEA 348 EUR
APOSPC405658-1G
3-Bromo-5-(pentafluorothio)benzyl bromide 97%
3-Bromo-5-(pentafluorothio)benzyl bromide
Formel: C₇H₅Br₂F₅S
Molecular Weight: 375,98 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16652484
CAS-Nummer: 1240257-14-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR