3-Methyl-5-(pentafluorosulphur)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC405663-1G
APOSPC405663-1GEA
422
EUR
APOSPC405663-1G
3-Methyl-5-(pentafluorosulphur)benzoic acid 97%
3-Methyl-5-(pentafluorosulphur)benzoic acid
Formel:
C₈H₇F₅O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD16652478 CAS-Nummer: 1448317-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...