3-Fluoro-5-(pentafluorothio)benzylamine 98%

Lieferant: Apollo Scientific

PC405665-1G
APOSPC405665-1GEA 348 EUR
APOSPC405665-1G
3-Fluoro-5-(pentafluorothio)benzylamine 98%
3-Fluoro-5-(pentafluorothio)benzylamine
Formel: C₇H₇F₆NS
Molecular Weight: 251,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16652463
CAS-Nummer: 1240257-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR