2-Chlor-4-(pentafluorthio)anilin
Lieferant: Apollo Scientific
|
Achtung
|
PC405667-1G
PC405667-5G
PC405667-10G
APOSPC405667-1GEA
0
EUR
APOSPC405667-1G
APOSPC405667-5G
APOSPC405667-10G
2-Chlor-4-(pentafluorthio)anilin
2-Chlor-4-(pentafluorthio)anilin
Formel:
C₆H₅ClF₅NS Schmelzpunkt: 56…58 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD20731080 CAS-Nummer: 165114-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...