2-Fluor-4-(pentafluorthio)anilin 97%

Lieferant: Apollo Scientific
Achtung

PC405669-1G PC405669-100MG
APOSPC405669-1GEA 575 EUR
APOSPC405669-1G APOSPC405669-100MG
2-Fluor-4-(pentafluorthio)anilin 97%
2-Fluor-4-(pentafluorthio)anilin
Formel: C₆H₅F₆NS
Lagertemperatur: Raumtemperatur
MDL: MFCD16652511
CAS-Nummer: 1240257-25-3
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR